missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene oxide, 97%

Product Code. 10591191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10591191 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10591191 Leverandør Thermo Scientific Acros Leverandørnr. 140140250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 503-30-0 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Tekniske data

CAS 503-30-0
Color Colorless
Density 0.8900g/mL
Boiling Point 50°C
Flash Point <1°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H6O
Refractive Index 1.3920 to 1.394
MDL Number MFCD00005167
Quantity 25 mL
Beilstein 17, 6
Fieser 01,1232
Merck Index 15, 9886
Specific Gravity 0.89
Synonym trimethylene oxide, 1,3-propylene oxide, 1,3-epoxypropane, oxacyclobutane, cyclooxabutane, trimethylenoxid, oxetan, 1,3-trimethylene oxide, propane, 1,3-epoxy, trimethylenoxid german
InChI Key AHHWIHXENZJRFG-UHFFFAOYSA-N
SMILES C1COC1
IUPAC Name oxetane
Molecular Weight (g/mol) 58.08
PubChem CID 10423
ChEBI CHEBI:30965
Formula Weight 58.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Trimethylene oxide, 97%
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.