missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene oxide, 97%

Product Code. 11457117
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457117 100 g 100g
11437117 5 g 5g
11447117 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457117 Supplier Thermo Scientific Alfa Aesar Supplier No. A14452.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 503-30-0 | C3H6O | 58.08 g/mol

Trimethylene oxide useful for the synthesis of 3-substituted propanols by reaction with Grignards or organolithiums in the presence of CuBr.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylene oxide useful for the synthesis of 3-substituted propanols by reaction with Grignards or organolithiums in the presence of CuBr.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is hygroscopic in nature and sensitive to air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503-30-0
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005167
InChI Key AHHWIHXENZJRFG-UHFFFAOYSA-N
Synonym trimethylene oxide, 1,3-propylene oxide, 1,3-epoxypropane, oxacyclobutane, cyclooxabutane, trimethylenoxid, oxetan, 1,3-trimethylene oxide, propane, 1,3-epoxy, trimethylenoxid german
PubChem CID 10423
ChEBI CHEBI:30965
IUPAC Name oxetane
SMILES C1COC1

Specifications

Melting Point -97°C
Density 0.891
Boiling Point 49°C to 50°C
Flash Point −28°C (−18°F)
Refractive Index 1.393
Quantity 100 g
UN Number UN1993
Beilstein 102382
Merck Index 14,9715
Solubility Information Soluble in water.
Formula Weight 58.08
Percent Purity 97%
Chemical Name or Material Trimethylene oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.