missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 1,3,5-benzenetricarboxylate, 99%, Thermo Scientific Chemicals

Product Code. 10001750
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2672-58-4
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.22
MDL Number MFCD00008434
InChI Key RGCHNYAILFZUPL-UHFFFAOYSA-N
Synonym trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
PubChem CID 75881
IUPAC Name trimethyl benzene-1,3,5-tricarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC

Specifications

Melting Point 144°C to 147°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CO2CH3)3
Quantity 100 g
Beilstein 09, 979
Formula Weight 252.22
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Trimethyl 1,3,5-benzenetricarboxylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'