missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylboroxin, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | Trimethylboroxin |
| CAS | 823-96-1 |
| Molecular Formula | C3H9B3O3 |
| Quantity | 2.5 g |
| Synonym | 2,4,6-Trimethyl-boroxin, Trimethyl-boroxin, 2,4,6-Trimethyl-1,3,5,2,4,6-trioxatriborinane, 2,4,6-Trimethylboroxine, Methaneboronic anhydride |
| InChI Formula | InChI=1S/C3H9B3O3/c1-4-7-5(2)9-6(3)8-4/h1-3H3 |
| SMILES | CB(OB(C)O1)OB1C |
| IUPAC Name | 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane |
| Molecular Weight (g/mol) | 125.53 |
| Formula Weight | 126.0831 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction