missing translation for 'onlineSavingsMsg'
Learn More

Trimethylboroxin (50% Solution in THF), TRC

Product Code. 30449444
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30449444 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30449444

Brand: Toronto Research Chemicals TRCT7961485G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 823-96-1
Molecular Formula C3H9B3O3
Quantity 5 g
Synonym 2,4,6-Trimethyl-boroxin, Trimethyl-boroxin, 2,4,6-Trimethyl-1,3,5,2,4,6-trioxatriborinane, 2,4,6-Trimethylboroxine, Methaneboronic anhydride
InChI Formula InChI=1S/C3H9B3O3/c1-4-7-5(2)9-6(3)8-4/h1-3H3
SMILES CB(OB(C)O1)OB1C
IUPAC Name 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol) 125.53
Formula Weight 126.0831
Concentration or Composition (by Analyte or Components) 50%
Chemical Name or Material Trimethylboroxin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.