missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine hydrochloride, 98%

Product Code. 11351687
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11351687

Brand: Thermo Scientific Alfa Aesar L04718.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 593-81-7 | C3H10ClN | 95.57 g/mol

Trimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.

Solubility
Soluble in water (737 mg/ml at 20°C), chloroform (moderately), and alcohol. Insoluble in ether.

Notes
Hygroscopic. Store under dry inert gas and away from moisture. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-81-7
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012478
InChI Key SZYJELPVAFJOGJ-UHFFFAOYSA-N
Synonym trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride
PubChem CID 10313079
ChEBI CHEBI:64700
IUPAC Name N,N-dimethylmethanamine;hydrochloride
SMILES [H+].[Cl-].CN(C)C

Specifications

Melting Point ∽280°C (decomposition)
Quantity 50 g
Beilstein 3905063
Sensitivity Hygroscopic
Merck Index 14,9710
Solubility Information Soluble in water (737mg/ml at 20°C),chloroform (moderately),and alcohol. Insoluble in ether.
Formula Weight 95.57
Percent Purity 98%
Chemical Name or Material Trimethylamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.