missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine (~30 wt. % Solution in Ethanol), TRC

Product Code. 30453751
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
Unit Size:
10mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453751 25 mL 25mL
30456439 50 mL 50mL
30452314 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453751 Supplier Toronto Research Chemicals Supplier No. TRCT79167525ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylamine
CAS 75-50-3
Molecular Formula C3 H9 N
Quantity 25 mL
Synonym Trimethylamine (8CI), N,N-Dimethylmethanamine, N,N,N-Trimethylamine, N-Trimethylamine, Methanamine, N,N-dimethyl-
InChI Formula InChI=1S/C3H9N/c1-4(2)3/h1-3H3
SMILES CN(C)C
IUPAC Name N,N-dimethylmethanamine
Molecular Weight (g/mol) 59.11
Formula Weight 59.0735
Concentration or Composition (by Analyte or Components) 30 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.