missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, 1M soln. in THF

Product Code. 15475667
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475667 500 mL 500mL
15465667 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475667 Supplier Thermo Scientific Alfa Aesar Supplier No. H32899.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-50-3 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine complexes of alane as precursors for the low-pressure chemical vapor deposition of aluminum. Trimethylamine was found to be unsuitable as an agent for differentiating between Lewis and Brpinsted sites because it is dissociatively adsorbed and is, therefore useful for the perturbing effects. Choline derivative manufactured from TMA are used in medication of liver disorders. Used directly as acid binder in the manufacture of Sulpha Drugs. Animal/poultry feed additive like choline chloride is manufactured from TMA. Used in the manufacture of alkyl quaternary ammonium salts. To manufacture Ion exchange Resins based on chloromethylated polystyrene.

Solubility
Easily soluble in water, also soluble in most organic solvents (such as methanol and ethanol).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
IUPAC Name N,N-dimethylmethanamine
SMILES CN(C)C

Specifications

Density 0.85
Linear Formula (CH3)3N
Quantity 500 mL
UN Number UN1993
Sensitivity Hygroscopic
Solubility Information Easily soluble in water,also soluble in most organic solvents (such as methanol and ethanol).
Formula Weight 59.11
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Trimethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.