missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetonitrile, 98%

Product Code. 10309043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10309043 100 mL Glass bottle 100mL
10153380 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10309043 Supplier Thermo Scientific Acros Supplier No. 140031000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 630-18-2 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-18-2
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00001847
InChI Key JAMNHZBIQDNHMM-UHFFFAOYSA-N
Synonym pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
PubChem CID 12416
IUPAC Name 2,2-dimethylpropanenitrile
SMILES CC(C)(C)C#N

Specifications

Melting Point 15°C to 16°C
Color Colorless
Density 0.7500g/mL
Boiling Point 105°C to 106°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCN
Refractive Index 1.3765 to 1.3785
Quantity 100 mL
Beilstein 02, 320
Specific Gravity 0.75
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone and toluene
Formula Weight 83.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trimethylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.