missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetonitrile, 98+%

Product Code. 11429862
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429862 100 g 100g
11419862 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429862 Supplier Thermo Scientific Alfa Aesar Supplier No. A12011.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 630-18-2 | C5H9N | 83.134 g/mol

Trimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.

Solubility
Miscible with ethanol, acetone and toluene. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-18-2
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001847
InChI Key JAMNHZBIQDNHMM-UHFFFAOYSA-N
Synonym pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
PubChem CID 12416
IUPAC Name 2,2-dimethylpropanenitrile
SMILES CC(C)(C)C#N

Specifications

Melting Point 16°C to 18°C
Density 0.757
Boiling Point 105°C to 106°C
Flash Point 4°C (39°F)
Odor Odorless
Quantity 100 g
UN Number UN3273
Solubility Information Miscible with ethanol,acetone and toluene. Slightly miscible with water.
Formula Weight 83.13
Percent Purity ≥98%
Chemical Name or Material Trimethylacetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethylacetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: 2,2-dimethyl-Propanenitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.