missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetonitrile, 98%, Thermo Scientific Chemicals

Product Code. 10153380
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-18-2
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00001847
InChI Key JAMNHZBIQDNHMM-UHFFFAOYSA-N
Synonym pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
PubChem CID 12416
IUPAC Name 2,2-dimethylpropanenitrile
SMILES CC(C)(C)C#N

Specifications

Melting Point 15°C to 16°C
Color Colorless
Density 0.7500g/mL
Boiling Point 105°C to 106°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCN
Refractive Index 1.3765 to 1.3785
Quantity 25 mL
Beilstein 02, 320
Specific Gravity 0.75
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone and toluene
Formula Weight 83.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trimethylacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'