missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetonitrile, 98+%

Product Code. 11419862
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11419862

Brand: Thermo Scientific Alfa Aesar A12011.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 630-18-2 | C5H9N | 83.134 g/mol

Trimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.

Solubility
Miscible with ethanol, acetone and toluene. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-18-2
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001847
InChI Key JAMNHZBIQDNHMM-UHFFFAOYSA-N
Synonym pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
PubChem CID 12416
IUPAC Name 2,2-dimethylpropanenitrile
SMILES CC(C)(C)C#N

Specifications

Melting Point 16°C to 18°C
Density 0.757
Boiling Point 105°C to 106°C
Flash Point 4°C (39°F)
Odor Odorless
Quantity 25 g
UN Number UN3273
Solubility Information Miscible with ethanol,acetone and toluene. Slightly miscible with water.
Formula Weight 83.13
Percent Purity ≥98%
Chemical Name or Material Trimethylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.