missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic anhydride, 99%

Product Code. 11496136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496136 25 g 25g
11406146 100 g 100g
11416146 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496136 Supplier Thermo Scientific Alfa Aesar Supplier No. B22983.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1538-75-6 | C10H18O3 | 186.25 g/mol

Trimethylacetic anhydride is used as acylation and esterification reagents. It is involved in acylation and esterification reactions with anilines and phenols. It is also used in solid-phase oligonucleotide synthesis and in the kinetic resolution of racemic 2-hydroxy-gamma-butyrolactones with diphenylacetic acid. It is also employed in the production of cyano-4, N-tert-butyloxycarbonyl piperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetic anhydride is used as acylation and esterification reagents. It is involved in acylation and esterification reactions with anilines and phenols. It is also used in solid-phase oligonucleotide synthesis and in the kinetic resolution of racemic 2-hydroxy-gamma-butyrolactones with diphenylacetic acid. It is also employed in the production of cyano-4, N-tert-butyloxycarbonyl piperidine.

Solubility
Miscible with acetonitrile.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1538-75-6
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00008842
InChI Key PGZVFRAEAAXREB-UHFFFAOYSA-N
Synonym pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride
PubChem CID 15234
IUPAC Name 2,2-dimethylpropanoyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC(=O)C(C)(C)C

Specifications

Density 0.918
Boiling Point 193°C
Flash Point 63°C (145°F)
Odor Unpleasant
Linear Formula ((CH3)3CCO)2O
Refractive Index 1.409
Quantity 25 g
UN Number UN3265
Beilstein 386552
Sensitivity Moisture sensitive
Solubility Information Miscible with acetonitrile.
Formula Weight 186.25
Percent Purity 99%
Chemical Name or Material Trimethylacetic anhydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethylacetic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Trimethylacetic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.