missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic acid, 99%

Product Code. 11459682
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459682 500 g 500g
11469682 2500 g 2500g
11479682 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11459682 Supplier Thermo Scientific Alfa Aesar Supplier No. A10776.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-98-9 | C5H10O2 | 102.133 g/mol

Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

Solubility
Soluble in alcohol, ether. Slightly soluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-98-9
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004194
InChI Key IUGYQRQAERSCNH-UHFFFAOYSA-N
Synonym pivalic acid, trimethylacetic acid, neopentanoic acid, 2,2-dimethylpropionic acid, tert-pentanoic acid, propanoic acid, 2,2-dimethyl, versatic 5, kyselina pivalova, acetic acid, trimethyl, alpha,alpha-dimethylpropionic acid
PubChem CID 6417
ChEBI CHEBI:45133
IUPAC Name 2,2-dimethylpropanoic acid
SMILES CC(C)(C)C(=O)O

Specifications

Melting Point 31°C to 35°C
Density 0.91
Boiling Point 163°C to 165°C
Flash Point 63°C (145°F)
Odor Pungent
Quantity 500 g
UN Number UN3261
Beilstein 969480
Merck Index 14,7511
Solubility Information Soluble in alcohol, ether; Slightly soluble in water
Formula Weight 102.13
Percent Purity 99%
Chemical Name or Material Trimethylacetic acid
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P235-Keep cool.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.