missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Thiophosphate, TRC

Product Code. 30450191
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450191 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450191 Supplier Toronto Research Chemicals Supplier No. TRCT7976051G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O,O-Trimethylthiophosphate
CAS 152-18-1
Molecular Formula C3 H9 O3 P S
Quantity 1 g
Synonym O,O,O-Trimethylphosphorthioate, Trimethoxy(thioxo)-lambda5-phosphane, Phosphorothioic acid','O,O,O-trimethyl ester (8CI','9CI','ACI), 7900-HC, HC 7900, Methyl phosphorothioate ((MeO)3PS), O,O,O-Trimethyl phosphorothioate, O,O,O-Trimethyl phosphorothionate, O,O,O-Trimethyl thiophosphate, SD 4741, Trimethoxyphosphine sulfide, Trimethyl thionophosphate
InChI Formula InChI=1S/C3H9O3PS/c1-4-7(8,5-2)6-3/h1-3H3
SMILES COP(=S)(OC)OC
IUPAC Name trimethoxy(sulfanylidene)-λ^{5}-phosphane
Molecular Weight (g/mol) 156.14
Formula Weight 156.001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.