missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl thiophosphate

Product Code. 11387268
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387268 5 g 5g
11397268 1 g 1g
11377268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387268 Supplier Thermo Scientific Alfa Aesar Supplier No. 030395.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 152-18-1 | C3H9O3PS | 156.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-18-1
Molecular Formula C3H9O3PS
Molecular Weight (g/mol) 156.136
MDL Number MFCD00014888
InChI Key XWSLYQXUTWUIKM-UHFFFAOYSA-N
Synonym trimethyl thiophosphate, trimethylthiophosphate, o,o,o-trimethyl phosphorothioate, o,o,o-trimethyl thiophosphate, trimethylthiofosfat, o,o,o-trimethylthiofosfat, trimethylthiofosfat czech, ccris 5569, phosphorothioic acid, o,o,o-trimethyl ester, o,o,o-trimethylthiofosfat czech
PubChem CID 9038
IUPAC Name trimethoxy(sulfanylidene)-$l^{5}-phosphane
SMILES COP(=S)(OC)OC

Specifications

Color White
Odor Sulphurous
Quantity 5 g
UN Number UN3278
Formula Weight 156.15
Physical Form Liquid
Chemical Name or Material Trimethyl thiophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.