missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl-tetradecylammonium Chloride, TRC

Product Code. 30453146
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453146 Supplier Toronto Research Chemicals Supplier No. TRCT798338500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetradecyltrimethylammonium Chloride
CAS 4574-04-3
Molecular Formula C17 H38 N . Cl
Quantity 500 mg
Synonym 1-Tetradecanaminium','N,N,N-trimethyl-','chloride (9CI), Ammonium','trimethyltetradecyl-','chloride (8CI), Trimethyltetradecylammonium chloride (6CI,7CI), C 14TAC, C14TAC, FSM 24, MTAC, Myristyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, TTAC, Tetradecyltrimethylammonium chloride, Trimethylmyristylammonium chloride, N,N,N-Trimethyl-1-tetradecanaminium chloride, Tetradecyltrimethylammonium Chloride
InChI Formula InChI=1S/C17H38N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name trimethyl(tetradecyl)azanium;chloride
Molecular Weight (g/mol) 291.94
Formula Weight 291.2693
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.