missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Phosphate, TRC

Product Code. 30451935
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451935 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451935 Supplier Toronto Research Chemicals Supplier No. TRCT796645100G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylphosphate
CAS 512-56-1
Purity Grade Notes GC
Molecular Formula C3 H9 O4 P
Quantity 100 g
Synonym Phosphoric acid','trimethyl ester (8CI','9CI','ACI), Methyl phosphate','(MeO)3PO (6CI), NSC 58985, TMP, TMPA, TMPO, Trimethoxyphosphine oxide, Trimethyl orthophosphate, Trimethyl phosphate
InChI Formula InChI=1S/C3H9O4P/c1-5-8(4,6-2)7-3/h1-3H3
SMILES COP(=O)(OC)OC
IUPAC Name trimethyl phosphate
Molecular Weight (g/mol) 140.07
Formula Weight 140.0238
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.