missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl phosphate, 99%, Thermo Scientific Chemicals

Product Code. 13448388
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-56-1
Molecular Formula C3H9O4P
Molecular Weight (g/mol) 140.08
MDL Number MFCD00008348
InChI Key WVLBCYQITXONBZ-UHFFFAOYSA-N
Synonym trimethylphosphate, phosphoric acid, trimethyl ester, trimethyl orthophosphate, tmpo, trimethoxyphosphine oxide, phosphoric acid trimethyl ester, o,o,o-trimethyl phosphate, trimethylfosfat, phosphate, trimethyl, trimethylfosfat czech
PubChem CID 10541
ChEBI CHEBI:46324
IUPAC Name trimethyl phosphate
SMILES COP(=O)(OC)OC

Specifications

Melting Point -46.0°C
Density 1.1970g/mL
Boiling Point 197.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)3P(O)
Refractive Index 1.3950 to 1.3970
Quantity 2.5 L
Beilstein 01,286
Fieser 05,716; 07,393
Specific Gravity 1.197
Solubility Information Solubility in water: 500g/L (25°C). Other solubilities: soluble in gasoline,soluble in alcohol,ether
Formula Weight 140.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethyl phosphate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl phosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H340-May cause genetic defects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'