missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(pentamethylcyclopentadienyl)titanium(IV), 97%

Product Code. 11300388
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11300388 2 g 2g
11390378 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11300388 Supplier Thermo Scientific Alfa Aesar Supplier No. 043759.04
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 107333-47-1 | C13H29Ti-5 | 233.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107333-47-1
Molecular Formula C13H29Ti-5
Molecular Weight (g/mol) 233.242
MDL Number MFCD00269851
InChI Key DVHMCUKDWMNRGX-UHFFFAOYSA-N
Synonym acmc-1bo0g, titanium,trimethyl 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
PubChem CID 57346580
IUPAC Name carbanide;cyclopenta-1,3-diene;titanium(4+)
SMILES [CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Ti+4]

Specifications

Packaging Ampule
Quantity 2 g
UN Number UN1325
Sensitivity Air, light, and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 228.22
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Trimethyl (pentamethylcyclopentadienyl)titanium (IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.