missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthovalerate, TRC

Product Code. 30450127
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450127 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450127 Supplier Toronto Research Chemicals Supplier No. TRCT79657550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyl Orthovalerate
CAS 13820-09-2
Molecular Formula C8H18O3
Quantity 50 g
Synonym 1,1,1-Trimethoxypentane, Orthovaleric Acid Trimethyl Ester, Trimethyl Orthopentanoate
InChI Formula InChI=1S/C8H18O3/c1-5-6-7-8(9-2,10-3)11-4/h5-7H2,1-4H3
SMILES CCCCC(OC)(OC)OC
IUPAC Name 1,1,1-trimethoxypentane
Molecular Weight (g/mol) 162.23
Formula Weight 162.1256
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.