missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthovalerate, 98%

Product Code. 11385649
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385649 100 g 100g
11365649 5 g 5g
11375649 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11385649 Supplier Thermo Scientific Alfa Aesar Supplier No. L20377.22
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13820-09-2 | C8H18O3 | 162.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-09-2
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00008481
InChI Key XUXVVQKJULMMKX-UHFFFAOYSA-N
Synonym trimethyl orthovalerate, pentane, 1,1,1-trimethoxy, unii-w0x9mom6vz, trimethylorthovalerate, orthovaleric acid, trimethyl ester, ortho-n-valeric acid trimethyl ester, w0x9mom6vz, trimethyl-2-valerate, trimethyl ortho-n-valerate, 1,1,1-trimethoxy-pentan
PubChem CID 83755
IUPAC Name 1,1,1-trimethoxypentane
SMILES CCCCC(OC)(OC)OC

Specifications

Boiling Point 164°C to 166°C
Flash Point 41°C (105°F)
Linear Formula CH3(CH2)3C(OCH3)3
Quantity 100 g
UN Number UN1993
Beilstein 1739200
Merck Index 14,9717
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material Trimethyl orthovalerate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.