missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthoformate, TRC

Product Code. 30450761
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450761 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450761 Supplier Toronto Research Chemicals Supplier No. TRCT796490250G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyl Orthoformate
CAS 149-73-5
Molecular Formula C4H10O3
Quantity 250 g
Synonym Methoxymethylal, Methyl Orthoformate, NSC 147479, Orthoformic Acid Methyl Ester, Perma-Flo OF, Trimethoxymethane
InChI Formula InChI=1S/C4H10O3/c1-5-4(6-2)7-3/h4H,1-3H3
SMILES COC(OC)OC
IUPAC Name trimethoxymethane
Molecular Weight (g/mol) 106.12
Formula Weight 106.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.