missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthobenzoate, 98%

Product Code. 11453993
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453993 10 g 10g
11463993 50 g 50g
11473993 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453993 Supplier Thermo Scientific Alfa Aesar Supplier No. A10270.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 707-07-3 | C10H14O3 | 182.219 g/mol

Trimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside. It reacts with indole to prepare indol-3-yl-phenyl-methanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside. It reacts with indole to prepare indol-3-yl-phenyl-methanone.

Solubility
Soluble in organic solvents.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-07-3
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00008474
InChI Key IECKAVQTURBPON-UHFFFAOYSA-N
Synonym trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate
PubChem CID 69720
IUPAC Name trimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)(OC)OC

Specifications

Density 1.06
Boiling Point 87°C to 88°C (7 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.489
Quantity 10 g
Beilstein 777623
Sensitivity Moisture sensitive
Solubility Information Soluble in organic solvents.
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material Trimethyl orthobenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.