missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthoacetate, 98+%

Product Code. 10308070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308070 100 mL Glass bottle 100mL
10647022 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10308070 Supplier Thermo Scientific Acros Supplier No. 218181000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1445-45-0 | C5H12O3 | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-45-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00008477
InChI Key HDPNBNXLBDFELL-UHFFFAOYSA-N
Synonym trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate
PubChem CID 15050
IUPAC Name 1,1,1-trimethoxyethane
SMILES CC(OC)(OC)OC

Specifications

Color Colorless to Brown
Density 0.9400g/mL
Boiling Point 107.0°C to 109.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(OCH3)3
Refractive Index 1.3870 to 1.3890
Quantity 100 mL
Beilstein 02, II, 128
Fieser 17,377; 01,834; 02,318
Specific Gravity 0.94
Solubility Information Solubility in water: hydrolysis. Other solubilities: forming methanol and acetic acid
Formula Weight 120.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Trimethyl orthoacetate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl orthoacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Trimethyl orthoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.