missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(methylcyclopentadienyl)platinum(IV)

Product Code. 15453017
Change view
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453017 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15453017 Supplier Thermo Scientific Alfa Aesar Supplier No. 098158.02
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 94442-22-5 | C9H16Pt | 319.31 g/mol

Trimethyl(methylcyclopentadienyl)platinum(IV) is used as an organometallic precursor for platinum. It is used in the microelectronics, catalysis, photonics and chemical sensing processes. It is used for the deposition of ultrathin metallic layers of platinum by the electron beam induced deposition process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 94442-22-5
Melting Point 28°C to 31°C
Color White
Boiling Point 23°C (0.053 mmHg)(sublimation)
Molecular Formula C9H16Pt
MDL Number MFCD00079665
Quantity 0.5 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
InChI Key AGQKROMSWCHOND-UHFFFAOYSA-N
SMILES C[Pt](C)C.Cc1cccc1
IUPAC Name trimethyl(2-methylcyclopenta-1,3-dien-1-yl)platinum
Molecular Weight (g/mol) 319.31
Formula Weight 319.32
Physical Form Pieces or Powder
Chemical Name or Material Trimethyl (methylcyclopentadienyl)platinum (IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.