missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl borate, 99.9995+% (metals basis)

Product Code. 11372588
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372588 50 g 50g
11392588 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372588 Supplier Thermo Scientific Alfa Aesar Supplier No. 022975.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-43-7 | C3H9BO3 | 103.912 g/mol

Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in the Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-43-7
Molecular Formula C3H9BO3
Molecular Weight (g/mol) 103.912
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Specifications

Melting Point -34°C
Color Colorless
Density 0.915 g/mL
Boiling Point 67°C to 69°C
Flash Point -8°C (17°F)
Assay Percent Range >99.9995% (metals basis)
Odor Odorless
Linear Formula (CH3O)3B
Refractive Index 1.361
Quantity 50 g
UN Number UN2416
Beilstein 1697939
Sensitivity Moisture sensitive
Merck Index 14,9712
Solubility Information Miscible with tetrahydrofuran,ether,isoporpylamine,hexane and methanol.
Formula Weight 103.92
Percent Purity ≥99.9995%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Trimethyl borate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.