missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 4-phosphonocrotonate, (E)+(Z), 90+%

Product Code. 11459563
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459563 250 g 250g
11439563 10 g 10g
11449563 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459563 Supplier Thermo Scientific Alfa Aesar Supplier No. A12399.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 86120-40-3 | C7H13O5P | 208.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86120-40-3
Molecular Formula C7H13O5P
Molecular Weight (g/mol) 208.15
MDL Number MFCD00051564
InChI Key PESRINKNQQZURC-SNAWJCMRSA-N
Synonym methyl 4-dimethoxyphosphoryl but-2-enoate, trimethyl 4-phosphonocrotonate, methyl 2e-4-dimethoxyphosphoryl but-2-enoate, methyl 4-dimethoxyphosphorylbut-2-enoate, trimethyl 4-phosphonocrotonate, e + z, methyl e-4-dimethoxyphosphorylbut-2-enoate, 4-dimethoxyphosphinyl crotonic acid methyl ester, e-4-dimethoxy-phosphoryl-but-2-enoic acid methyl ester
PubChem CID 5702556
IUPAC Name methyl (E)-4-dimethoxyphosphorylbut-2-enoate
SMILES COC(=O)\C=C\CP(=O)(OC)OC

Specifications

Boiling Point 123°C to 124°C (1 mmHg)
Quantity 250 g
Beilstein 4987133
Formula Weight 208.15
Percent Purity ≥90%
Chemical Name or Material Trimethyl 4-phosphonocrotonate, (E)+(Z)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.