missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 1,3,5-benzenetricarboxylate, 98%

Product Code. 11495007
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495007 25 g 25g
11485007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495007 Supplier Thermo Scientific Alfa Aesar Supplier No. B21926.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2672-58-4 | C12H12O6 | 252.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2672-58-4
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.222
MDL Number MFCD00008434
InChI Key RGCHNYAILFZUPL-UHFFFAOYSA-N
Synonym trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
PubChem CID 75881
IUPAC Name trimethyl benzene-1,3,5-tricarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC

Specifications

Melting Point 144°C to 147°C
Quantity 25 g
Beilstein 2219698
Formula Weight 252.22
Percent Purity 98%
Chemical Name or Material Trimethyl 1,3,5-benzenetricarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.