missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(pentamethylcyclopentadienyl)titanium(IV), 97+%

Product Code. 11387509
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387509 5 g 5g
11377509 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387509 Supplier Thermo Scientific Alfa Aesar Supplier No. H27885.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123927-75-3 | C13H24O3Ti+ | 276.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123927-75-3
Molecular Formula C13H24O3Ti+
Molecular Weight (g/mol) 276.199
MDL Number MFCD00269850
InChI Key GQQYDESAAOEXPK-UHFFFAOYSA-N
PubChem CID 17755674
IUPAC Name methanolate;1,2,3,4,5-pentamethylcyclopentane;titanium(4+)
SMILES C[C]1[C]([C]([C]([C]1C)C)C)C.C[O-].C[O-].C[O-].[Ti+4]

Specifications

Density 1.081
Boiling Point 285°C
Flash Point 61°C (142°F)
Refractive Index 1.554
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 276.21
Percent Purity ≥97%
Chemical Name or Material Trimethoxy(pentamethylcyclopentadienyl)titanium(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.