missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim, MedChemExpress

Product Code. 16450968
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450968 5 g 5g
16408518 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16450968 Supplier Medchem Express Supplier No. HYB0510
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trimethoprim is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. Trimethoprim is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. Trimethoprim has the potential for the research of urinary tract infections, Shigellosis and Pneumocystis pneumonia. Trimethoprim can inhibit infection of Influenza A virus in chick embryo when combinated with zinc.

Trimethoprim is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. Trimethoprim is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. Trimethoprim has the potential for the research of urinary tract infections, Shigellosis and Pneumocystis pneumonia. Trimethoprim can inhibit infection of Influenza A virus in chick embryo when combinated with zinc.

Specifications

Chemical Name or Material Trimethoprim
Physical Form Solid
CAS 738-70-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C14H18N4O3
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 50 mg/mL (172.22 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.31 mM; Need ultrasonic)
SMILES NC1=NC=C(CC2=CC(OC)=C(OC)C(OC)=C2)C(N)=N1
Molecular Weight (g/mol) 290.32
Formula Weight 290.32
Percent Purity 99.96%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.