missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoprim-d4 (Major), TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Trimethoprim-d3 |
| CAS | 1189923-38-3 |
| Molecular Formula | C14 D5 H15 N4 O3 |
| Quantity | 10 mg |
| Synonym | Trimethoprim D3 (4-methoxy D3) |
| InChI Formula | InChI=1S/C14H18N4O3.2H/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15;;/h5-7H,4H2,1-3H3,(H4,15,16,17,18);;/i3D3;2*1+1 |
| SMILES | [2H].[2H].[2H]C([2H])([2H])Oc1c(OC)cc(Cc2cnc(N)nc2N)cc1OC |
| IUPAC Name | 5-[[3,5-dimethoxy-4-(trideuteriomethoxy)phenyl]methyl]pyrimidine-2,4-diamine |
| Molecular Weight (g/mol) | 297.364 |
| Formula Weight | 297.185 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction