missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethobenzamide hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Description
Trimethobenzamide hydrochloride is a blocker of the D2 receptor. Trimethobenzamide is an antiemetic used to prevent nausea and vomiting.
Specifications
Specifications
| Chemical Name or Material | Trimethobenzamide hydrochloride |
| CAS | 554-92-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H29ClN2O5 |
| Quantity | 200 mg |
| Synonym | Ro 2-9578 |
| Solubility Information | DMSO : ≥ 100 mg/mL (235.34 mM) |
| SMILES | O=C(NCC1=CC=C(OCCN(C)C)C=C1)C2=CC(OC)=C(OC)C(OC)=C2.Cl |
| Molecular Weight (g/mol) | 424.92 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction