missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethobenzamide hydrochloride, MedChemExpress
Description
Trimethobenzamide hydrochloride is a blocker of the D2 receptor. Trimethobenzamide is an antiemetic used to prevent nausea and vomiting.
Specifications
Specifications
| Chemical Name or Material | Trimethobenzamide hydrochloride |
| CAS | 554-92-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H29ClN2O5 |
| Quantity | 500 mg |
| Synonym | Ro 2-9578 |
| Solubility Information | DMSO : ≥ 100 mg/mL (235.34 mM) |
| SMILES | O=C(NCC1=CC=C(OCCN(C)C)C=C1)C2=CC(OC)=C(OC)C(OC)=C2.Cl |
| Molecular Weight (g/mol) | 424.92 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?