missing translation for 'onlineSavingsMsg'
Learn More

Trimetazidine dihydrochloride, MedChemExpress

Product Code. 16434517
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434517 50 mg 50mg
16408648 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16434517 Supplier Medchem Express Supplier No. HYB0968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trimetazidine dihydrochloride is a selective long chain 3-ketoyl coenzyme A thiolase inhibitor with an IC50 of 75 nM, which can inhibit β-oxidation of free fatty acid (FFA).

Trimetazidine dihydrochloride is an effective antianginal agent and a cytoprotective drug, has anti-oxidant, anti-inflammatory, antinociceptive and gastroprotective properties. Trimetazidine dihydrochloride triggers autophagy. Trimetazidine dihydrochloride is also a 3-hydroxyacyl-CoA dehydrogenase (HADHA) inhibitor.

Specifications

Chemical Name or Material Trimetazidine dihydrochloride
CAS 13171-25-0
Color White
Purity Grade Notes Research
Molecular Formula C14H24Cl2N2O3
Quantity 50 mg
Solubility Information H2O : ≥ 100 mg/mL (294.76 mM) ∣DMSO : 25 mg/mL (73.69 mM; Need ultrasonic)
SMILES COC1=CC=C(CN2CCNCC2)C(OC)=C1OC.[H]Cl.[H]Cl
Molecular Weight (g/mol) 339.26
Formula Weight 339.26
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.