missing translation for 'onlineSavingsMsg'
Learn More

Trimesinic Acid, TRC

Product Code. 30451863
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451863 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451863 Supplier Toronto Research Chemicals Supplier No. TRCT7956455G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 554-95-0
Molecular Formula C9H6O6
Quantity 5 g
Synonym 1,3,5-Tricarboxybenzene, 5-Carboxyisophthalic Acid, NSC 3998
InChI Formula InChI=1S/C9H6O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H,(H,10,11)(H,12,13)(H,14,15)
SMILES OC(=O)c1cc(cc(c1)C(=O)O)C(=O)O
IUPAC Name benzene-1,3,5-tricarboxylic acid
Molecular Weight (g/mol) 210.14
Formula Weight 210.0164
Chemical Name or Material Trimesinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.