missing translation for 'onlineSavingsMsg'
Learn More

Trimellitic Anhydride, TRC

Product Code. 30457056
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
30457056 50 g 50g
30456476 10 g 10g
30450073 250 g -
3 options
This item is not returnable. View return policy

Product Code. 30457056

Brand: Toronto Research Chemicals TRCT79586550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-30-7
Formula Weight 192.0059
InChI Formula InChI=1S/C9H4O5/c10-7(11)4-1-2-5-6(3-4)9(13)14-8(5)12/h1-3H,(H,10,11)
IUPAC Name 1,3-dioxo-2-benzofuran-5-carboxylic acid
Molecular Formula C9 H4 O5
Molecular Weight (g/mol) 192.125
SMILES OC(=O)c1ccc2C(=O)OC(=O)c2c1
Synonym 5-Isobenzofurancarboxylic acid, 1,3-dihydro-1,3-dioxo-, 1,2,4-Benzenetricarboxylic acid, anhydride (7CI), 1,2,4-Benzenetricarboxylic acid, cyclic 1,2-anhydride (8CI), Trimellitic acid anhydride (6CI), 1,3-Dihydro-1,3-dioxo-5-isobenzofurancarboxylic acid, 1,2,4-Benzenetricarboxylic acid 1,2-anhydride, 1,2,4-Benzenetricarboxylic anhydride, 1,3-Dioxo-1,3-dihydro-2-benzofuran-5-carboxylic acid, 1,3-Dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, 1,3-Dioxo-5-phthalancarboxylic acid, 4-Carboxyphthalic anhydride, Anhydrotrimellitic acid, Aradur 3380-1, Benzene-1,3,4-tricarboxylic anhydride, CE-X 23 Medium, Epon 9150, NSC 60252, TMA, TMA (anhydride), Trimellitic acid cyclic 1,2-anhydride, Trimellitic anhydride
Chemical Name or Material Trimellitic acid 1,2-anhydride

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.