missing translation for 'onlineSavingsMsg'
Learn More

Trimellitic anhydride chloride, 98%

Product Code. p-7046456
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11373506

Brand: Thermo Scientific Alfa Aesar L14382.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1204-28-0 | C9H3ClO4 | 210.569 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1204-28-0
Molecular Formula C9H3ClO4
Molecular Weight (g/mol) 210.569
MDL Number MFCD00005924
InChI Key NJMOHBDCGXJLNJ-UHFFFAOYSA-N
Synonym trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride
PubChem CID 70998
IUPAC Name 1,3-dioxo-2-benzofuran-5-carbonyl chloride
SMILES C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O

Specifications

Melting Point 67°C to 69°C
Quantity 25 g
UN Number UN3261
Beilstein 610823
Sensitivity Moisture sensitive
Formula Weight 210.57
Percent Purity 98%
Chemical Name or Material Trimellitic anhydride chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.