missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimebutine, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Trimebutine |
| CAS | 39133-31-8 |
| Analyte or Component Names | Trimebutine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H29 N O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trimebutine, 3,4,5-Trimethoxybenzoic acid 2-(dimethylamino)-2-phenylbutyl ester, (+/-)-Trimebutine, BRL-9476 |
| InChI Formula | InChI=1S/C22H29NO5/c1-7-22(23(2)3,17-11-9-8-10-12-17)15-28-21(24)16-13-18(25-4)20(27-6)19(14-16)26-5/h8-14H,7,15H2,1-6H3 |
| Solution Type | Neat |
| SMILES | CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(N(C)C)c2ccccc2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction