missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM9 Antibody (OTI2D6), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74616APCCY7
This item is not returnable.
View return policy
Description
TRIM9 Monoclonal specifically detects TRIM9 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| TRIM9 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TRIM9 | |
| Recombinant protein fragment corresponding to amino acids 284-669 of human TRIM9 (NP_443210) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI2D6 | |
| Flow Cytometry | |
| EC 6.3.2.-, KIAA0282homolog of rat RING finger Spring, RNF91RING finger protein 91, SPRINGE3 ubiquitin-protein ligase TRIM9, tripartite motif containing 9, tripartite motif-containing 9, Tripartite motif-containing protein 9 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 114088.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction