missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM8 Recombinant Rabbit Monoclonal Antibody (HL2797)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556723
This item is not returnable.
View return policy
Description
TRIM8 Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to nuclear bodies. Its structure is similar to some tumor suppressor proteins and its gene maps to a locus thought to contain tumor suppressor genes.
Specifications
| TRIM8 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9BZR9 | |
| TRIM8 | |
| Recombinant fragment of human GERP. | |
| 100 μL | |
| Primary | |
| Human, Rat, Feline | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2797 | |
| Unconjugated | |
| TRIM8 | |
| AA408830; BB006755; E3 ubiquitin-protein ligase TRIM8; GERP; glioblastoma expressed ring finger protein; glioblastoma-expressed RING finger protein; Probable E3 ubiquitin-protein ligase TRIM8; ring finger protein 27; RING-type E3 ubiquitin transferase TRIM8; Rnf27; TRIM8; tripartite motif containing 8; tripartite motif protein 8; tripartite motif protein TRIM8; tripartite motif-containing 8; tripartite motif-containing protein 8; tripartite motif-containing protein 8; probable E3 ubiquitin-protein ligase TRIM8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 101082095, 688785, 81603 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction