missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27463-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to nuclear bodies. Its structure is similar to some tumor suppressor proteins and its gene maps to a locus thought to contain tumor suppressor genes.Specifications
| TRIM8 | |
| Polyclonal | |
| Unconjugated | |
| TRIM8 | |
| GERP, RING finger protein 27, RNF27, TRIM8, tripartite motif containing 8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 81603 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BZR9 | |
| TRIM8 | |
| TRIM8 Fusion Protein Ag26517 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction