missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRIM8 293T Cell Transient Overexpression Lysate

Product Code. 18444799 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
This item is not returnable. View return policy

Product Code. 18444799

Brand: Novus Biologicals™ H00081603T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Research Category Zinc Finger
Target TRIM8
Synonym EC 6.3.2.-, GERP, GERPRING finger protein 27 glioblastoma expressed ring finger protein, Glioblastoma-expressed RING finger protein, RNF27probable E3 ubiquitin-protein ligase TRIM8, tripartite motif containing 8, tripartite motif protein TRIM8, tripartite motif-containing 8, Tripartite motif-containing protein 8
Gene Symbol TRIM8
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.