missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM72 Recombinant Rabbit Monoclonal Antibody (HL1855)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547201
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Muscle-specific protein that plays a central role in cell membrane repair by nucleating the assembly of the repair machinery at injury sites. Specifically binds phosphatidylserine. Acts as a sensor of oxidation: upon membrane damage, entry of extracellular oxidative environment results in disulfide bond formation and homooligomerization at the injury site. This oligomerization acts as a nucleation site for recruitment of TRIM72-containing vesicles to the injury site, leading to membrane patch formation. Probably acts upstream of the Ca(2+)-dependent membrane resealing process. Required for transport of DYSF to sites of cell injury during repair patch formation. Regulates membrane budding and exocytosis. May be involved in the regulation of the mobility of KCNB1-containing endocytic vesicles.
Specifications
| TRIM72 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| A0JPQ4, Q1XH17, Q6ZMU5 | |
| TRIM72 | |
| Recombinant fragment of human TRIM72. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1855 | |
| Unconjugated | |
| TRIM72 | |
| BC067209; mg53; mitsugumin 53; mitsugumin-53; RGD1562778; TRIM72; trim72 {ECO:0000250; tripartite motif containing 72; tripartite motif containing 72, E3 ubiquitin protein ligase; tripartite motif-containing 72; tripartite motif-containing protein 72; UniProtKB:Q8N4X6} | |
| Rabbit | |
| Protein A | |
| RUO | |
| 365377, 434246, 493829 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction