missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM7 Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26285-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1, a B-box type 2, and a coiled-coil region. The protein localizes to both the nucleus and the cytoplasm, and may represent a participant in the initiation of glycogen synthesis. Multiple transcript variants have been found for this gene, and some of them encode the same isoform.Specifications
| TRIM7 | |
| Polyclonal | |
| Unconjugated | |
| Trim7 | |
| Glycogenin interacting protein, GNIP, RING finger protein 90, RNF90, TRIM7, tripartite motif containing 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 94089 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q923T7 | |
| Trim7 | |
| TRIM7 Fusion Protein Ag23453 | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction