missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM69 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12951-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
| TRIM69 | |
| Polyclonal | |
| Unconjugated | |
| TRIM69 | |
| HSD 34, HSD34, RING finger protein 36, RNF36, Trif, TRIM69, tripartite motif containing 69 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140691, 311373, 70928 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5BK82, Q80X56, Q86WT6 | |
| Trim69 | |
| TRIM69 Fusion Protein Ag4074 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction