missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM61 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561884
This item is not returnable.
View return policy
Description
Immunogen sequence: KLEEYNAPWK ERVELIEKVI TMQTRKSLEL KKKMESPSV Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 36%.
RNF35 belongs to the TRIM/RBCC family.
Specifications
| TRIM61 | |
| Polyclonal | |
| Unconjugated | |
| TRIM61 | |
| 2czf61; AU018704; C85201; E330039K03Rik; putative tripartite motif-containing protein 61; RING finger protein 35; RNF35; TRIM61; tripartite motif containing 61; tripartite motif-containing 61 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 391712 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5EBN2 | |
| TRIM61 | |
| Recombinant protein corresponding to Human TRIM61. Recombinant protein control fragment (Product #RP-100772). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction