missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM60 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557407
This item is not returnable.
View return policy
Description
Immunogen sequence: SYHREILEGS LEPLRNNIER VEKVIILQGS KSVELKKKVE YKREEINSEF EQIRLFLQNE QEMILRQIQD EEMNILAKLN ENLVELSDYV STL Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 32%.
The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Pseudogenes of this gene are located on more than six chromosomes including chromosome 4. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2013].
Specifications
| TRIM60 | |
| Polyclonal | |
| Unconjugated | |
| TRIM60 | |
| 2czf45; RING finger protein 129; ring finger protein 33; RNF129; Rnf33; TRIM60; tripartite motif containing 60; tripartite motif-containing 60; tripartite motif-containing protein 60 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 166655 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q495X7 | |
| TRIM60 | |
| Recombinant protein corresponding to Human TRIM60. Recombinant protein control fragment (Product #RP-96745). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction