missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM60 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49338
This item is not returnable.
View return policy
Description
TRIM60 Polyclonal antibody specifically detects TRIM60 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TRIM60 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FLJ35882, RING finger protein 129MGC119325, RING finger protein 33, RNF129, RNF33, tripartite motif containing 60, tripartite motif-containing 60, tripartite motif-containing protein 60 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VGNKPKWILGVCQDCLLRNWQDQPSVLGGFWAIGRYMKSGYVASGPKTTQLLPVVKPSK | |
| 0.1 mL | |
| Zinc Finger | |
| 166655 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction