missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM58 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572893
This item is not returnable.
View return policy
Description
TRIM58 Polyclonal Antibody for Western Blot, ELISA
E3 ubiquitin ligase induced during late erythropoiesis. Directly binds and ubiquitinates the intermediate chain of the microtubule motor dynein (DYNC1LI1/DYNC1LI2), stimulating the degradation of the dynein holoprotein complex. May participate in the erythroblast enucleation process through regulation of nuclear polarization.
Specifications
| TRIM58 | |
| Polyclonal | |
| Unconjugated | |
| TRIM58 | |
| BIA2; E3 ubiquitin-protein ligase TRIM58; Gm16; Gm1783; Protein BIA2; RGD1566174; RING-type E3 ubiquitin transferase TRIM58; TRIM58; tripartite motif containing 58; tripartite motif-containing 58; tripartite motif-containing protein 58 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 216781, 25893, 303167 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q5NCC9, Q8NG06 | |
| TRIM58 | |
| TRIM58 antibody for a peptide corresponding to 17 amino acids near the carboxy terminus of human TRIM58. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction